missing translation for 'onlineSavingsMsg'
Learn More

MK-2894, MedChemExpress

Product Code. 16456677
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456677

Brand: Medchem Express HY104135mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-2894 is a potent, selective, orally active and high affinity (Ki=0.56 nM) full antagonist against E prostanoid receptor 4 (EP4 receptor) (IC50=2.5 nM). MK-2894 possesses potent anti-inflammatory activity in animal models of pain/inflammation and can be used for the research of arthritis.

MK-2894 is a potent, selective, orally active and high affinity (Ki=0.56 nM) full antagonist against E prostanoid receptor 4 (EP4 receptor) (IC50=2.5 nM). MK-2894 possesses potent anti-inflammatory activity in animal models of pain/inflammation and can be used for the research of arthritis.

Specifications

Chemical Name or Material MK-2894
Physical Form Solid
CAS 1006036-87-8
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H22F3NO3S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (105.59 mM)
SMILES O=C(O)C1=CC=C(C2(NC(C3=C(C)SC(C)=C3CC4=CC=C(C(F)(F)F)C=C4)=O)CC2)C=C1
Molecular Weight (g/mol) 473.51
Formula Weight 473.51
Percent Purity 95.14%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.