missing translation for 'onlineSavingsMsg'
Learn More

MK 212 hydrochloride, Tocris Bioscience™

Product Code. 13438183
Encompass
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT2C agonist

5-HT receptor agonist; displays selectivity for 5-HT2C over 5-HT2A (IC50 values are 0.028 and 0.42 μM for human 5-HT2C and 5-HT2A receptors expressed in HEK293 cells respectively).

Specifications

CAS 64022-27-1
Molecular Formula C8H12Cl2N4
MDL Number MFCD04975444
Quantity 10mg
Synonym 2-chloro-6-1-piperazinyl pyrazine, 6-chloro-2-1-piperazinyl pyrazine, 2-chloro-6-piperazin-1-yl pyrazine, pyrazine, 2-chloro-6-1-piperazinyl, tocris-0941, biomol-nt_000130, d02esw, gtpl165, 1-6-chloro-2-pyrazinyl piperazine, 6-chloro-2-1-piperazinyl-pyrazine
InChI Key PFIZGLUIYAZQFU-UHFFFAOYSA-N
SMILES [H+].[Cl-].ClC1=CN=CC(=N1)N1CCNCC1
IUPAC Name hydrogen 2-chloro-6-(piperazin-1-yl)pyrazine chloride
Molecular Weight (g/mol) 235.11
PubChem CID 107992
Formula Weight 235.12
Percent Purity >99%
Chemical Name or Material MK 212 hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'