missing translation for 'onlineSavingsMsg'
Learn More

MK-1064, MedChemExpress

Product Code. p-200101436
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16450428 5 mg 5mg
16427756 100 mg 100mg
16423897 50 mg 50mg
16417968 10 mg 10mg
16413716 25 mg 25mg
16401899 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16450428 Supplier Medchem Express Supplier No. HY19914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-1064 is a selective and orally active OX2R antagonist (Ki: 0.5 nM, IC50: 18 nM). MK-1064 promotes sleep in vivo. MK-1064 can be used in the research of insomnia.

MK-1064 is a selective and orally active OX2R antagonist (Ki: 0.5 nM, IC50: 18 nM). MK-1064 promotes sleep in vivo. MK-1064 can be used in the research of insomnia.

Specifications

Chemical Name or Material MK-1064
CAS 1207253-08-4
Color White
Purity Grade Notes Research
Molecular Formula C24H20ClN5O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (108.25 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=CC(Cl)=CN=C2)=CN=C1C3=NC=CC=C3)NCC4=NC(OC)=C(OC)C=C4
Molecular Weight (g/mol) 461.9
Formula Weight 461.9
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.