missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-1064, MedChemExpress
Description
MK-1064 is a selective and orally active OX2R antagonist (Ki: 0.5 nM, IC50: 18 nM). MK-1064 promotes sleep in vivo. MK-1064 can be used in the research of insomnia.
Specifications
Specifications
| Chemical Name or Material | MK-1064 |
| CAS | 1207253-08-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H20ClN5O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50 mg/mL (108.25 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(C2=CC(Cl)=CN=C2)=CN=C1C3=NC=CC=C3)NCC4=NC(OC)=C(OC)C=C4 |
| Molecular Weight (g/mol) | 461.9 |
| Formula Weight | 461.9 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?