missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MK 0677
GREENER_CHOICE

Product Code. p-7110798
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18747428

Brand: Tocris Bioscience™ 5272/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity ghrelin receptor agonist

MK 0677 is a high affinity ghrelin receptor agonist (pKi = 8.14). Growth hormone (GH) secretagog; stimulates GH release from rat pituitary cells in vitro (EC50 = 1.3 nM) and enhances GH plasma levels in vivo. Also attenuates isoproterenol -induced lipolysis in rat adipocytes in vitro. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-N-[(1 R)-2-[1,2-dihydro-1-(methylsulfonyl)spiro[3 H-indole-3,4 '-piperidin]-1 '-yl]-2-oxo-1-[(phenylmethoxy)methyl]ethyl]-2-methylpropanamide methanesulfonate
CAS 159752-10-0
Synonym Ibutamoren mesylate, L-163,191
Quantity 10 mg
Target Ghrelin Receptor Agonists
Molecular Formula C27H36N4O5S.CH3SO3H
Formula Weight 624.77
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.