missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MK 0677

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MK 0677 is a high affinity ghrelin receptor agonist (pKi = 8.14). Growth hormone (GH) secretagog; stimulates GH release from rat pituitary cells in vitro (EC50 = 1.3 nM) and enhances GH plasma levels in vivo. Also attenuates isoproterenol -induced lipolysis in rat adipocytes in vitro. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 2-Amino-N-[(1 R)-2-[1,2-dihydro-1-(methylsulfonyl)spiro[3 H-indole-3,4 '-piperidin]-1 '-yl]-2-oxo-1-[(phenylmethoxy)methyl]ethyl]-2-methylpropanamide methanesulfonate |
| CAS | 159752-10-0 |
| Synonym | Ibutamoren mesylate, L-163,191 |
| Quantity | 50 mg |
| Target | Ghrelin Receptor Agonists |
| Molecular Formula | C27H36N4O5S.CH3SO3H |
| Formula Weight | 624.77 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction