missing translation for 'onlineSavingsMsg'
Learn More

MK-0249, MedChemExpress

Product Code. 16412348
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16412348

Brand: Medchem Express HYU00076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-0249 (Compound 1) is a potent, selective and orally active histamine H3 receptor antagonist, with an IC50 of 1.7 nM for human H3.

MK-0249 (Compound 1) is a potent, selective and orally active histamine H3 receptor antagonist, with an IC50 of 1.7 nM for human H3.

Specifications

Chemical Name or Material MK-0249
CAS 862309-06-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H24F3N3O2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (115.89 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(OCCCN3CCCC3)C=C2)C(C)=NC4=C1C(C(F)(F)F)=CC=C4
Molecular Weight (g/mol) 431.45
Formula Weight 431.45
Percent Purity 96.94%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.