missing translation for 'onlineSavingsMsg'
Learn More

Mizolastine, MedChemExpress

Product Code. 16428146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428146 100 mg 100mg
16472046 500 mg 500mg
16471386 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428146 Supplier Medchem Express Supplier No. HYB0164

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mizolastine is a histamine H1-receptor antagonist with IC50 of 47 nM used in the treatment of hay fever (seasonal allergic rhinitis), hives and other allergic reactions.

Mizolastine is a histamine H1-receptor antagonist with IC50 of 47 nM used in the treatment of hay fever (seasonal allergic rhinitis), hives and other allergic reactions.

Specifications

Chemical Name or Material Mizolastine
CAS 108612-45-9
Color White
Purity Grade Notes Research
Molecular Formula C24H25FN6O
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (57.80 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NC(N(C2CCN(C3=NC4=CC=CC=C4N3CC5=CC=C(F)C=C5)CC2)C)=NC=C1
Molecular Weight (g/mol) 432.49
Formula Weight 432.49
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.