missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miz-1/ZBTB17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£231.00 - £470.00
Specifications
| Antigen | Miz-1/ZBTB17 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407020
|
Novus Biologicals
NBP1-81048-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257335
|
Novus Biologicals
NBP1-81048 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Miz-1/ZBTB17 Polyclonal specifically detects Miz-1/ZBTB17 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Miz-1/ZBTB17 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Miz-1, MIZ1, Myc-interacting zinc finger protein 1, pHZ-67, zinc finger and BTB domain containing 17, zinc finger and BTB domain-containing protein 17, Zinc finger protein 151, zinc finger protein 151 (pHZ-67), Zinc finger protein 60MIZ-1, ZNF151Myc-interacting Zn finger protein-1, ZNF60 | |
| ZBTB17 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q13105, Q13105, Q13105, Q13105 | |
| 7709 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSPENVDDVLAVATFLQMQDIITACHALKSLAEPATSPGGNAEALATEGGDKRAKEEKVATSTLSRLEQAGRSTPIGPSRDLKEERGGQAQSAASGAEQTEKADAPREPPPVELKPDPTSGMAAAEAEAALSESSEQEMEVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title