missing translation for 'onlineSavingsMsg'
Learn More

Mixture of (R,S)-N-Nitroso Anabasine-d4, rac N'-Nitrosonornicotine-d4, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone-d4 and (S)-N-Nitroso Anatabine-d4 (1.0mg/mL in Acetonitrile), TRC

Product Code. 30401509
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401509 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30401509 Supplier Toronto Research Chemicals Supplier No. TRCKIT21551X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-N-Nitrosoanabasine pyridinyl-d4∣(S)-N-Nitrosoanatabine 2,4,5,6 D4∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4)∣N-Nitrosonornicotine-ketoneD4(pyridylD4)
CAS 1020719-68-9, 1426174-82-4, 66148-19-4, 764661-24-7
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (RS)-N-Nitrosoanabasine pyridinyl-d4 1.0mg/mL∣(S)-N-Nitrosoanatabine 2,4,5,6 D4 1.0mg/mL∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4) 1.0mg/mL∣N-Nitrosonornicotine-ketoneD4(pyridylD4) 1.0mg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.