missing translation for 'onlineSavingsMsg'
Learn More

Mixture of Ibuprofen (1.0 mg/ml) and Paracetamol (1.0 mg/ml) in Methanol, Mikromol

Product Code. 30457421
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30457421 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457421 Supplier Mikromol Supplier No. MM0002.M1L11000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mixture of Ibuprofen (1.0 mg/ml) and Paracetamol (1.0 mg/ml) in Methanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen∣Paracetamol
CAS 15687-27-1, 103-90-2
Analyte or Component Names Ibuprofen∣Paracetamol
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solution Type Mixture
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1 mg/mL (Ibuprofen), 1 mg/mL (Paracetamol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.