missing translation for 'onlineSavingsMsg'
Learn More

Mixture of Glycerol 1-phosphate and Glycerol 2-phosphate Hydrate (Technical Grade), TRC

Product Code. 30393891
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393891 5 g 5g
30393396 1 g 1g
30391568 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393891 Supplier Toronto Research Chemicals Supplier No. TRCG2527105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mixture of Glycerol 1-phosphate and Glycerol 2-phosphate Hydrate
Analyte or Component Names Mixture of Glycerol 1-phosphate and Glycerol 2-phosphate Hydrate
Shipping Condition Room Temperature
Molecular Formula C3H9Na2O6P • xH2O
Synonym Sodium 2-Hydroxy-1-(hydroxymethyl)ethyl Phosphate Hydrate (2:1)
InChI Formula InChI=1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2
Solution Type Neat
SMILES OCC(OP(O[Na])(O[Na])=O)CO.[H2O]
IUPAC Name sodium 1,3-dihydroxypropan-2-yl phosphate hydrate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 234.06
Quantity 5 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.