Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC), 100L, for HyPerforma™ Mixtainer™ Systems
Utilize these bioprocess containers with a magnetic stir bar for mixing applications in bioproduction.
Brand: Thermo Scientific™ SH30687.05
Additional Details : Weight : 0.45000kg
Description
Provide a closed system with single-use contact surfaces and an integrated magnetic stir bar with these bioprocess containers for Thermo Scientific™ HyPerforma™ Mixtainer™ Systems.
Optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products available for sterile liquid/liquid or powder/liquid mixing.Mixtainer BioProcess Container — The Mixtainer BPC is supplied sterilized by irradiation. It is constructed of Thermo Scientific CX5-14 film and contains a magnetic stir-bar located in a plastic dish welded in the bottom of the BPC. The dish contains a locking ring to ensure the containment of the stir-bar during shipping, handling and operation. A powder port option is available on one of the standard configurations. Aseptic liquid transfers are facilitated by connectors and tubing attached to ports at the top of the BPC
Conical Drum — The Mixtainer system utilizes a polyethylene drum with a special enlarged opening at the bottom to accommodate the stir-bar dish
Magnetic Stirring Plate —A commercially available standard unit with the power and variable speed capability to provide a range of mixing
Support Plate — The support plate provides the right fit to ensure properfunctioning
Dolly —The dolly is designed to interface between the drum and the magnetic stirring plate to create the vertical and concentric alignment necessary for operation. It also provides mobility
- Available in 50, 100, and 200L unit volumes
- Recommended for applications where the BPC contents are post filtered
Recommended for:
- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Specifications
100 L | |
CX5-14 | |
Packages placed in strong cardboard cartons with product and lot identification | |
Gamma Irradiated | |
6.3 mm (0.25 in.) Insert luer lock, tube length: 91 cm (36 in.) | |
9.53 mm (0.38 in.) Quick Connect, body tube length: 91 cm (36 in.) | |
Fewer resources used and less waste |
Mixtainer BioProcess Container, 100 L | |
Powder addition port 38.1 mm (1.5 in.) tri-clamp port, Top Ports: 2 x 0.25 in., 2 x 0.38 in. | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
9.5 mm (0.38 in.) Quick Connect Body with 16.5 cm (6.5 in.) dip tube, tube length: 91 cm (36 in.) | |
6.3 mm (0.25 in.) Insert luer lock, tube length: 91 cm (36 in.) | |
HyPerforma™, Mixtainer™ |