missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIXL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567818
This item is not returnable.
View return policy
Description
Predicted to react with Human samples.
MIXL1 is a novel human Mix-like homeobox gene. In normal hematopoiesis, its expression appears to be restricted to immature B- and T-lymphoid cells.
Specifications
| MIXL1 | |
| Polyclonal | |
| Unconjugated | |
| MIXL1 | |
| hMix; Homeobox protein MIXL1; homeodomain protein MIX; MILD1; MIX; Mix paired-like homeobox; Mix paired-like homeobox 1; mix.1 homeobox-like protein; Mix1 homeobox-like 1 (Xenopus laevis); MIX1 homeobox-like protein 1; MIXL; Mixl1; Mix-like homeobox protein 1; Mm1; mMix; Mml | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27217, 83881 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H2W2, Q9WUI0 | |
| MIXL1 | |
| Synthesized peptide derived from human MIXL1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction