Learn More
Thermo Scientific™ Mixed Multi Mini Water Softener
Product Code. 15256343 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
LabTower EDI
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15256343 LabTower EDI Each
1 options
Product Code. 15256343
Brand: Thermo Scientific™ 50130297
Only null left
Add to basket
Add to basket
Treat unsoftened tap water with the Thermo Scientific™ Mixed Multi Mini water softener, required if feed water silt density index (SDI) is greater than 3.
Treat unsoftened tap water or a TIS of more than 10ppm with the Thermo Scientific™ Mixed Multi Mini Water Softener. For use with Thermo Scientific™ Barnstead™ water purification systems, it is required if feed water silt density index (SDI) is greater than 3.
Required for use: Softener salt, hardness detection kit and 5μm filter with carbon. This softener is ideal for use with the Barnstead LabTower™ EDI water purification system.
TRUSTED_SUSTAINABILITY
Specifications
Description Mix Multi Mini Water softener
For Use With (Equipment) LabTower EDI
Voltage 230 V