missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed indicator solution, For determination of water hardness, according to Cooper, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
1 L
1.5 L
6 x 250 mL
6 x 1 L
Unit Size:
1 L
1.5 L
250mL
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Mixed indicator solution |
| Quantity | 6 x 250 mL |
| Flash Point | 11°C (51.8°F) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction