missing translation for 'onlineSavingsMsg'
Learn More

Mixed indicator solution, For determination of water hardness, according to Cooper, Honeywell Fluka™

Product Code. 15635050 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
1 L
1.5 L
6 x 250 mL
6 x 1 L
Unit Size:
1 L
1.5 L
250mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15635050

Brand: Honeywell-Fluka 360231L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for determination of water hardness, according to Cooper

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mixed indicator solution
Quantity 1 L
Flash Point 11°C (51.8°F)
compliance-icons compliance-icons
Product Identifier
  • Mixed indicator solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.