missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MF-Millipore™ Membrane Filter, MCE, 0.45 μm
Mixed cellulose esters (MCE) membrane, hydrophilic
Brand: Merck HAWP05000
172.42 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
4 L/min/ cm2 | |
White | |
MCE | |
HAWP | |
Used in analytical/research applications | |
100Pack | |
75°C | |
Filter Membranes | |
50 mm |
≥1.8 bar | |
Membrane Filter | |
0.45 μm | |
4 L/min/ cm2 (Air), 60 mL/min/ cm2 (Water) | |
79% | |
Plain | |
150 μm | |
50 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Indicazioni di pericolo
Consigli di prudenza
Informazioni supplementari
- MF-Millipore Membrane Filter, 0.45
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable solid Category 1
Indicazioni di pericolo
- H228-Flammable solid.
- H412-Harmful to aquatic life with long lasting effects.
Consigli di prudenza
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Informazioni supplementari
- MIXTURE LIST-Contains : Octylphenol polyethoxyethanol