missing translation for 'onlineSavingsMsg'
Learn More

Mixed Acid, 10% (v/v) Hydrofluoric Acid/10% (v/v) Nitric Acid, Ricca Chemical

Product Code. 31940464
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940464 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940464 Supplier Ricca Chemical Company Supplier No. R51513001A
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Chemical Name or Material 10% (v/v) Nitric acid-10% (v/v) hydrofluoric acid mixtures
Packaging Natural Poly Bottle
CAS 7664-39-3, 7697-37-2, 7732-18-5
Physical Form Liquid
Color Colorless
pH < 2
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Analytical Volumetric Solution
Quantity 1 L
Percent Purity 0.1
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.