missing translation for 'onlineSavingsMsg'
Learn More

Mitragynine (100μg/mL in Methanol), TRC

Product Code. 30398604
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398604 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398604 Supplier Toronto Research Chemicals Supplier No. TRCKIT02901X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4098-40-2
Molecular Formula C23 H30 N2 O4
Quantity 1 mL
Synonym Indolo[2,3-a]quinolizine-2-acetic acid, 3-ethyl-1,2,3,4,6,7,12,12b-octahydro-8-methoxy-α-(methoxymethylene)-, methyl ester, (αE,2S,3S,12bS)-, Corynan-16-carboxylic acid, 16,17-didehydro-9,17-dimethoxy-, methyl ester, (16E,20β)-, Corynantheidine, 9-methoxy- (7CI), Mitragynine (6CI), (-)-Mitragynine, 9-Methoxycorynantheidine, Indolo[2,3-a]quinolizine-2-acetic acid, 3-ethyl-1,2,3,4,6,7,12,12b-octahydro-8-methoxy-α-(methoxymethylene)-, methyl ester, [2S-[2α(E),3α,12bβ]]-, Mitragynin, Kratom
InChI Formula InChI=1S/C23H30N2O4/c1-5-14-12-25-10-9-15-21-18(7-6-8-20(21)28-3)24-22(15)19(25)11-16(14)17(13-27-2)23(26)29-4/h6-8,13-14,16,19,24H,5,9-12H2,1-4H3/b17-13+/t14-,16+,19+/m1/s1
SMILES CC[C@@H]1CN2CCc3c([nH]c4cccc(OC)c34)[C@@H]2C[C@@H]1\C(=C/OC)\C(=O)OC
IUPAC Name methyl (E)-2-[(2S,3S,12bS)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
Molecular Weight (g/mol) 398.5
Formula Weight 398.2206
Concentration or Composition (by Analyte or Components) Mitragynine 100 μg/mL
Chemical Name or Material Mitragynine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.