missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitoxantrone, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Mitoxantrone is a potent topoisomerase II inhibitor. Mitoxantrone also inhibits protein kinase C (PKC) activity with an IC50 of 8.5 μM. Mitoxantrone induces apoptosis of B-CLL (B-chronic lymphocytic leukaemia) cells. Mitoxantrone shows antitumor activity. Mitoxantrone also has anti-orthopoxvirus activity with EC50s of 0.25 μM and and 0.8 μM for cowpox and monkeypox, respectively.
Specifications
Specifications
| Chemical Name or Material | Mitoxantrone |
| CAS | 65271-80-9 |
| Color | Black |
| Purity Grade Notes | Research |
| Molecular Formula | C22H28N4O6 |
| Quantity | 50 mg |
| Synonym | Mitozantrone NSC 301739 |
| Solubility Information | DMSO : 62.5 mg/mL (140.61 mM; Need ultrasonic) |
| SMILES | O=C1C2=C(C(NCCNCCO)=CC=C2NCCNCCO)C(C3=C(O)C=CC(O)=C13)=O |
| Molecular Weight (g/mol) | 444.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction