missing translation for 'onlineSavingsMsg'
Learn More

Mitotane, TRC

Product Code. 30135259
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135259 10 g 10g
30134176 50 mg 50mg
17992015 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135259 Supplier Toronto Research Chemicals Supplier No. TRCM37250010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-19-0
Molecular Formula C14 H10 Cl4
Molecular Weight (g/mol) 320.04
Synonym Ethane, 1,1-dichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)- (6CI,7CI,8CI), 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene, 1,1-Dichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)ethane, 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane, 1-(2'-Chlorophenyl)-1-(4'-chlorophenyl)-2,2-dichloroethane, 1-(o-Chlorophenyl)-1-(p-chlorophenyl)-2,2-dichloroethane, 2,4'-Dichlorodiphenyldichloroethane, 2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethane, 2-(p-Chlorophenyl)-2-(o-chlorophenyl)-1,1-dichloroethane, CB 313, Chloditan, Chlodithane, Lysodren, Mitotan, Mitotane, NSC 38721, Opeprim, o,p'-DDD, o,p'-Dichlorodiphenyldichloroethane, o,p'-TDE
IUPAC Name 1-chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene
SMILES ClC(Cl)C(c1ccc(Cl)cc1)c2ccccc2Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H10Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8,13-14H
Formula Weight 317.9537
Chemical Name or Material 2,4'-DDD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.