missing translation for 'onlineSavingsMsg'
Learn More

Mitoquinone mesylate, MedChemExpress

Product Code. 16452526
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452526 25 mg 25mg
16495517 5 mg 5mg
16402788 10 mg 10mg
16449476 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452526 Supplier Medchem Express Supplier No. HY100116A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage.

Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage.

Specifications

Chemical Name or Material Mitoquinone mesylate
CAS 845959-50-4
Color Brown
Purity Grade Notes Research
Molecular Formula C38H47O7PS
Quantity 25 mg
Synonym MitoQ mesylateMitoQ10 mesylate
Solubility Information DMSO : 50 mg/mL (73.66 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (12.27 mM; ultrasonic and warming and heat to 60°C)
SMILES [O-]S(=O)(C)=O.O=C(C(CCCCCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=C4C)C(OC)=C(OC)C4=O
Molecular Weight (g/mol) 678.81
Formula Weight 678.81
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.