missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitoquinone mesylate, MedChemExpress
Description
Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage.
Specifications
Specifications
| Chemical Name or Material | Mitoquinone mesylate |
| CAS | 845959-50-4 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C38H47O7PS |
| Quantity | 5 mg |
| Synonym | MitoQ mesylateMitoQ10 mesylate |
| Solubility Information | DMSO : 50 mg/mL (73.66 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (12.27 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | [O-]S(=O)(C)=O.O=C(C(CCCCCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=C4C)C(OC)=C(OC)C4=O |
| Molecular Weight (g/mol) | 678.81 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?