missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mitomycin C
GREENER_CHOICE

Product Code. p-200085189
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
This item is not returnable. View return policy

Product Code. 18792554

Brand: Tocris Bioscience™ 3258/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DNA cross-linking antitumor agent

Mitomycin C is an antibiotic and antitumor agent. Covalently binds DNA forming intra- and interstrand crosslinks. Inhibits DNA synthesis. Also used for MEF/STO feeder layer preparation in stem cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5

Specifications

Quantity 10 mg
Formula Weight 334.33
Product Type Mitomycin C
Purity >98%
Molecular Weight (g/mol) 334.332
Chemical Name or Material Mitomycin C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.