missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mitomycin C

Product Code. 17190192
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190192 Supplier STEMCELL Technologies Supplier No. 73274
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic; Double-stranded DNA alkylating agent

  • Mitomycin C is an antibiotic which acts as a double-stranded DNA alkylating agent
  • It covalently crosslinks DNA, inhibiting DNA synthesis and cell proliferation
  • It acts by way of reductive activation either through low pH or NAD(P)H:quinone oxidoreductase (DT-diaphorase) or NADH cytochrome c reductase (Mao et al.; Cummings et al.)
  • Mitotically inactivates mouse embryonic fibroblasts (MEFs) for use as feeder cell layers in embryonic stem cell co-culture systems (Bryja et al.)
  • Selectively inhibits DNA synthesis and mutagenesis, stimulates genetic recombination, chromosome breakage and sister chromatid exchange, and induces DNA repair (Tomasz)

Specifications

CAS 50-07-7
Quantity 10 mg
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.