missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitofusin 2 Antibody (S153-5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42188AF647
This item is not returnable.
View return policy
Description
Mitofusin 2 Monoclonal specifically detects Mitofusin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Mitofusin 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MFN2 | |
| Fusion protein amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2. Rat: 97% identity (226/231 amino acids identical). Human: 92% identity (214/231 amino acids identical) ∼55% identity with Mitofusin-1. | |
| 0.1 mL | |
| 9927 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S153-5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CMT2A, CMT2A2, CPRP1mitochondrial assembly regulatory factor, EC 3.6.5, EC 3.6.5.-, HSG, KIAA0214hyperplasia suppressor, MARF, mitofusin 2, mitofusin-2, Transmembrane GTPase MFN2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Approx 55% identity with Mitofusin-1.Detects approx 90kDa. No cross-reactivity against Mitofusin-1. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction