missing translation for 'onlineSavingsMsg'
Learn More

Mitofusin 2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18403336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403336 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403336 Supplier Novus Biologicals Supplier No. NBP266383DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mitofusin 2 Polyclonal antibody specifically detects Mitofusin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitofusin 2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CMT2A, CMT2A2, CPRP1mitochondrial assembly regulatory factor, EC 3.6.5, EC 3.6.5.-, HSG, KIAA0214hyperplasia suppressor, MARF, mitofusin 2, mitofusin-2, Transmembrane GTPase MFN2
Host Species Rabbit
Immunogen Partial recombinant human MFN2 protein (amino acids 364-599). [O95140]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 9927
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.