missing translation for 'onlineSavingsMsg'
Learn More

Mitofilin Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18346876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346876 20 μL 20µL
18302344 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346876 Supplier Novus Biologicals Supplier No. NBP30334620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mitofilin Polyclonal specifically detects Mitofilin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitofilin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp779P1653, inner membrane protein, mitochondrial, MGC111146, Mic 60, Mic60, MICOS Complex Subunit MIC60, mitochondrial (mitofilin), mitofilin, motor protein, p87/89, proliferation-inducing gene 4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-270 of human Mitofilin (NP_006830.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10989
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.