missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitocondrial Translational Initiation Factor 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38558-20ul
This item is not returnable.
View return policy
Description
Mitocondrial Translational Initiation Factor 3 Polyclonal antibody specifically detects Mitocondrial Translational Initiation Factor 3 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Mitocondrial Translational Initiation Factor 3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human Mitocondrial Translational Initiation Factor 3 (NP_690876.3).,, Sequence:, MAALFLKRLTLQTVKSENSCIRCFGKHILQKTAPAQLSPIASAPRLSFLIHAKAFSTAEDTQNEGKKTKKNKTAFSNVGRKISQRVIHLFDEKGNDLGNMHRANVIRLMDERDLRLVQRNTSTEPAEYQLMTGLQILQERQRLREMEKANPKTGPTLRKELILSSNIGQHDLDTKTKQIQQWIKKKHLVQITIKKGKNVDVSENEMEEIFHQILQTMPGIATFSSRPQAVQGGKALMCVLRAFSKNEEKAYKETQETQERDTLNKDHGNDKESNVLHQ | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 219402 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction