missing translation for 'onlineSavingsMsg'
Learn More

Mitochonic acid 5, MedChemExpress

Product Code. 16490797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490797 50 mg 50mg
16487207 5 mg 5mg
16485356 100 mg 100mg
16474488 10 mg 10mg
16423116 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490797 Supplier Medchem Express Supplier No. HY111536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage. Mitochonic acid 5 modulates mitochondrial ATP synthesis.

Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage. Mitochonic acid 5 modulates mitochondrial ATP synthesis.

Specifications

Chemical Name or Material Mitochonic acid 5
CAS 1354707-41-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H13F2NO3
Quantity 50 mg
Synonym MA-5
Solubility Information DMSO : ≥ 106.66 mg/mL (323.90 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C(CC(C1=CC=C(F)C=C1F)=O)C2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 329.3
Formula Weight 329.3
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.