missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial Ribosomal Protein S18C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83211-0.1ML
This item is not returnable.
View return policy
Description
Mitochondrial Ribosomal Protein S18C Polyclonal specifically detects Mitochondrial Ribosomal Protein S18C in Human samples. It is validated for Western Blot.
Specifications
| Mitochondrial Ribosomal Protein S18C | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51023 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 28S ribosomal protein S18c, mitochondrial, CGI-134, mitochondrial ribosomal protein S18-1, mitochondrial ribosomal protein S18C, MRP-S18-1, MRPS18-1, MRPS18-1mitochondrial, mrps18-c, MRP-S18-c, MRPS18C mitochondrial ribosomal protein S18C, S18mt-c | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human Mitochondrial Ribosomal Protein S18C | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction