missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mitochondrial ribosomal protein L4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85292-0.1ML
This item is not returnable.
View return policy
Description
mitochondrial ribosomal protein L4 Polyclonal specifically detects mitochondrial ribosomal protein L4 in Human samples. It is validated for Western Blot.
Specifications
| mitochondrial ribosomal protein L4 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CGI-28, L4mt, MGC16367, MGC2681, mitochondrial ribosomal protein L4,39S ribosomal protein L4, mitochondrial, MRP-L4 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human mitochondrial ribosomal protein L4 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51073 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction