missing translation for 'onlineSavingsMsg'
Learn More

mitochondrial ribosomal protein L4 Antibody (4B1), Novus Biologicals™

Product Code. 18355999 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355999 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355999 Supplier Novus Biologicals Supplier No. H00051073M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

mitochondrial ribosomal protein L4 Monoclonal antibody specifically detects mitochondrial ribosomal protein L4 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen mitochondrial ribosomal protein L4
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00756
Gene Alias CGI-28, L4mt, MGC16367, MGC2681, mitochondrial ribosomal protein L4,39S ribosomal protein L4, mitochondrial, MRP-L4
Host Species Mouse
Immunogen MRPL4 (AAH00756, 1 a.a. ~ 263 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLQFVRAGARAWLRPTGSQGLSSLAEEAARATENPEQVASEGLPEPVLRKVELPVPTHRRPVQAWVESLRGFEQERVGLADLHPDVFATAPRLDILHQVAMWQKNFKRISYAKTKTRAEVRGGGRKPWPQKGTGRARHGSIRSPLWRGGGVAHGPRGPTSYYYMLPMKVRALGLKVALTVKLAQDDLHIMDSLELPTGDPQYLTELAHYRRWGDSVLLVDLTHEEMPQSIVEATSRLKTFNLIPAVGEQRAQAPRVRMCRLRC
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51073
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.