missing translation for 'onlineSavingsMsg'
Learn More

mitochondrial ribosomal protein L30 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18393885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393885 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393885 Supplier Novus Biologicals Supplier No. NBP305921UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mitochondrial ribosomal protein L30 Polyclonal specifically detects mitochondrial ribosomal protein L30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mitochondrial ribosomal protein L30
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 39S ribosomal protein L30, mitochondrial, L28mt, L30mt, mitochondrial ribosomal protein L30, MRP-L28FLJ44438, MRPL28MGC3314,39S ribosomal protein L28, mitochondrial, MRP-L30, RPML28MGC24095
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human mitochondrial ribosomal protein L30 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51263
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.