missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial ribosomal protein L11 Antibody (OTI1E5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72707APC
This item is not returnable.
View return policy
Description
Mitochondrial ribosomal protein L11 Monoclonal antibody specifically detects Mitochondrial ribosomal protein L11 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Mitochondrial ribosomal protein L11 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1E5 | |
| Western Blot, Immunohistochemistry | |
| L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial | |
| Full length human recombinant protein of human MRPL11(NP_057134) produced in E.coli. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 65003 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction