missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial ribosomal protein L11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06019-50ul
This item is not returnable.
View return policy
Description
Mitochondrial ribosomal protein L11 Polyclonal antibody specifically detects Mitochondrial ribosomal protein L11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Mitochondrial ribosomal protein L11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human Mitochondrial ribosomal protein L11 fragment. | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 65003 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction