missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial-processing peptidase subunit beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | Mitochondrial-processing peptidase subunit beta |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314946
|
Novus Biologicals
NBP3-03227-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18385806
|
Novus Biologicals
NBP3-03227-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mitochondrial-processing peptidase subunit beta Polyclonal specifically detects Mitochondrial-processing peptidase subunit beta in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Mitochondrial-processing peptidase subunit beta | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9512 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Beta-MPP, EC 3.4.24.64, mitochondrial processing peptidase beta subunit, mitochondrial-processing peptidase subunit beta, MPPBMPP11, MPPP52, P-52, peptidase (mitochondrial processing) beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-380 of human Mitochondrial-processing peptidase subunit beta (NP_004270.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title