missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mitochondrial Hydroxyl Radical Detection Assay Kit, Fluorescent, 200 Test

Product Code. 32109873 Shop All Abcam Products
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109873 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109873 Supplier Abcam Supplier No. ab219931200Test
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescence-based assay kit for detecting hydroxyl radicals in suspension and adherent cell samples.

  • Detection method: Fluorescent
  • Sample types: Suspension cells, Adherent cells
  • Compatible instruments: Microplate reader, Fluorescence microscope
  • Cited in: > 5 publications (7 total)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Detection Location In-Cell Detection
For Use With (Equipment) Microplate Reader, Fluorescence Microscope
Format Kit
No. of Reactions 200 Tests
Target Molecule Hydroxyl Radical
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 200 Tests
For Use With (Application) Hydroxyl radical detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.