missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mitochondria Monoclonal Antibody (113-1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547691

Product Code. 17856558

  • £362.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specific for endogenous levels of the ~60 kDa human mitochondria. The antibody works on acetone fixed and 2% paraformaldehyde fixed cells. The antibody also works for immunohistochemistry on 2% paraformaldehyde fixed (5-10 minutes) cryostat sections.

Antibodies that recognize subcellular organelles or compartments of human cells are useful in identification of these organelles in cells, tissues, and biochemical preparations. Anti-mitochondria antibodies can be used to stain mitochondria in cell/tissue preparations and can be used as a marker in subcellular fractions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mitochondria
Monoclonal
2.0 mg/mL
PBS with no preservative
Q01844, Q61545
Human cell homogenate.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
113-1
Unconjugated
EWSR1
Mouse
Protein A
RUO
14030, 2130, 609786
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.