missing translation for 'onlineSavingsMsg'
Learn More

Mitochondria Antibody (MTC754), Novus Biologicals™

Mouse Monoclonal Antibody

£227.00 - £373.00

Specifications

Antigen Mitochondria
Clone MTC754
Concentration 0.2 mg/mL
Dilution Western Blot 0.25-0.5 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18434712
View Documents
Novus Biologicals
NBP2-32874-0.1mg
0.1 mg
£373.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18478981
View Documents
Novus Biologicals
NBP2-32874-0.02mg
0.02 mg
£227.00
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mitochondria Monoclonal antibody specifically detects Mitochondria in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Mitochondria
0.2 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cellular Markers, Mitochondrial Markers
10 mM PBS with 0.05% BSA
IgG1 κ
Protein A or G purified
MTC754
Western Blot 0.25-0.5 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Monoclonal
Purified
RUO
Human
Mitochondrial fraction of HeLa cells
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.