missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondria Antibody (AE-1) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48282
This item is not returnable.
View return policy
Description
Mitochondria Monoclonal specifically detects Mitochondria in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Mitochondria | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Semi-purified mitochondrial preparation | |
| Protein A or G purified | |
| RUO | |
| monoclonal antibody MTC02 recognizes a 60kDa antigen associated with the mitochondria in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. monoclonal antibody MTC02 recognizes an antigen associated with the mitochondria in human cells only. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. monoclonal antibody MTC02 is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells. | |
| Store at 4C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AE-1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Mouse | |
| 60 kDa | |
| 7 mL | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction