missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MitoBrilliant Live 646

Product Code. 30040308
Click to view available options
Quantity:
50 μg (Trial size)
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 30040308

Brand: Tocris Bioscience™ 7417/50U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red fluorescent mitochondrial stain for live cells, Δψm dependent

MitoBrilliant probes have been developed to overcome some common limitations encountered with standard mitochondrial trackers, offering clearer answers to scientific questions. They are next-generation fluorescent stains for the localization and tracking of mitochondria in both live and fixed cells. The range harnesses Janelia Fluor® dye technology, conferring some of the properties that make these widely used dyes, into mitochondrial stains.

The MitoBrilliant™ Live dyes accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner. Upon loss of the mitochondrial membrane potential, the dyes disperse, and so provide a dynamic assessment of the mitochondrial membrane potential in live cells. Two dyes are available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).

A universal stain for mitochondria in both live and fixed cells is also available: MitoBrilliant™ 646 .

Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.

MitoBrilliant is a trademark of Bio-Techne Corp.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg (Trial size)
Target Mitochondria Dyes and Probes
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.