missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mithramycin A, 90%, from Streptomyces argillaceus

Product Code. 10432971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Packaging:
Glass Bottle
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432971 5 mg Glass Bottle 5mg
10599213 1 mg Glass Bottle 1mg
10608424 25 mg Glass Bottle 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432971 Supplier Thermo Scientific Chemicals Supplier No. 415090050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24
Molecular Weight (g/mol) 1085.156
MDL Number MFCD00135618
InChI Key CFCUWKMKBJTWLW-ZUZMCERRSA-N
PubChem CID 122172942
IUPAC Name (2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dih
SMILES CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)O

Specifications

Type Mithramycin A
Physical Form Powder
CAS Min % 88.0
CAS Max % 100.0
Melting Point 180.0°C to 183.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Brown to Red
Quantity 5 mg
Assay Percent Range 90%
Merck Index 15, 7652
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in lower alcohols, acetone, ethyl acetate, moderatly soluble in chloroform, slightly soluble in ether, benzene
Formula Weight 1085.16
Percent Purity 90%
Chemical Name or Material Mithramycin A, 90%
Show More Show Less
compliance-icons
Product Identifier
  • Mithramycin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.