Learn More
Invitrogen™ MiTF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588618
Description
Immunogen sequence: MLEMLEYNHY QVQTHLENPT KYHIQQAQRQ QVKQYLSTTL ANKHANQVLS LPCPNQPGDH VMPPVPGSSA PNSPMAMLTL NSNCEKEGFY KFEEQNRAES ECPGMNTHSR ASCMQMDDVI DDIISLESSY NEEILGLMDP ALQMANTLPV SGNLIDLYGN QGLPPPGLTI SNSCPANLPN IKRELTESEA RALAKERQKK DNHNLIERRR RFNINDRIKE LGTLIPKSND PDMRWNKGTI LKASVDYIRK LQREQQRAKE LENRQKKLEH ANRHLLLRIQ; Positive Samples: Jurkat, HeLa, K-562, Mouse kidney; Cellular Location: Nucleus.
Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, while the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.
Specifications
| MiTF | |
| Polyclonal | |
| Unconjugated | |
| MITF | |
| BCC2; Bhlhe32; black eyed white; bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; mi; microphtalmia-associated transcription factor; microphthalmia; microphthalmia associated transcription factor isoform M-; microphthalmia transcription factor; microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; MITF-M-; transcription factor, isoform 7; vit; Vitiligo; Wh; WS2; WS2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17342, 4286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O75030, Q08874 | |
| MITF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human MITF (NP_9378011). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.