missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MITF Antibody (MITF/915), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11593CL1
This item is not returnable.
View return policy
Description
MITF Monoclonal antibody specifically detects MITF in Canine,Human,Mouse (Negative),Rat (Negative) samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence
Specifications
| MITF | |
| Monoclonal | |
| CoraFluor 1 | |
| BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
| Recombinant full-length human MITF protein (Uniprot: O75030) | |
| 0.1 mL | |
| Primary | |
| Canine, Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| MITF/915 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4286 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction