missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MITF Polyclonal antibody specifically detects MITF in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | MITF |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant human MITF, Val119-Lys289, Accession # O75030 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?