missing translation for 'onlineSavingsMsg'
Learn More

MITF Antibody (21D1418) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£183.00 - £417.00

Specifications

Antigen MITF
Clone 21D1418
Concentration 1.0 mg/ml
Dilution Western Blot 0.1 - 1.0 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 μg/mL, Immunohistochemistry-Paraffin 1:200
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18070534
View Documents
Novus Biologicals
NB100-56561SS
0.025 mg
£183.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18729543
View Documents
Novus Biologicals
NB100-56561
0.1 mg
£417.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MITF Monoclonal specifically detects MITF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MITF
1.0 mg/ml
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse, Rat
O75030
4286
A synthetic peptide mapping to the 300-350 a.a. region of accession number O75030.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
21D1418
Western Blot 0.1 - 1.0 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 μg/mL, Immunohistochemistry-Paraffin 1:200
Monoclonal
Purified
RUO
PBS with 0.02% Sodium Azide
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A
MITF
IgG1
Protein G purified
Mitf produces, by alternative splicing, 10 different transcripts, all with introns, putatively encoding 10 different protein isoforms. Therefore, observed molecular weights may vary depending on MITF isoforms expressed. Users are encouraged to utilize the peptide immunogen sequence to gain further information about the isoforms recognized by the antibody.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.