missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MITD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | MITD1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MITD1 Polyclonal specifically detects MITD1 in Human samples. It is validated for Western Blot.Specifications
| MITD1 | |
| Polyclonal | |
| Rabbit | |
| Q8WV92 | |
| 129531 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| LOC129531, MIT domain-containing protein 1, MIT, microtubule interacting and transport, domain containing 1 | |
| MITD1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title