Learn More
Emma Safety Footwear Mitchel Safety Shoes
Product Code. 15988241
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15988241 39 1 pair
15998241 40 1 pair
15908251 41 1 pair
15918251 42 1 pair
15928251 43 1 pair
15938251 44 1 pair
15948251 45 1 pair
15958251 46 1 pair
15968251 47 1 pair
15978251 48 1 pair
15988251 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15988241 Supplier Emma Safety Footwear Supplier No. MM15484839
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction with extra wear-resistant coating
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and cold insulated
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Mitchel
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 39
Toe Material Steel